Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 3/4" ID x 1/2" ID; 1/Each
Representative image only.
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 3/4" ID x 1/2" ID; 1/Each
Eldon James | Mfr: C12-8SS| Item: 5012770
Excellent for high temperature and harsh physical environments
₹3,700.00INR / Each
₹3,700.00INR / Each
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
  • 3/4" hose barb x 1/2" hose barb
More About this Item
These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. Use the straight reducers to connect two different size tubing. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialStainless Steel
  • Sterilization MethodE-beam/gamma irradiation, EtO, or autoclave up to 134°C
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 1) (in)3/4
  • Barb Size / Tubing Id (Port 2) (in)1/2
  • Flow PatternStraight
  • ColorMetallic
  • Regulatory ApprovalsNon-DEHP;RoHS;Non-Phthalate
  • DescriptionHose Barb Fitting, Straight Reducer, Stainless Steel, 3/4" ID x 1/2" ID; 1/EA