Eldex Optos Series Maintenance Kit for Pumps with 316 SS Cylinder and 1/4" Dia Piston
Representative image only.
Eldex Optos Series Maintenance Kit for Pumps with 316 SS Cylinder and 1/4" Dia Piston
Eldex | Mfr: 5922| Item: 4853193
Extend the life of your pump by keeping it well maintained
₹153,000.00INR / Each
₹153,000.00INR / Each
Product Description
Includes
Two GFP piston seals, two check valve cartridges, retainer, filter, and tools.
Key Features
  • For pumps with 316 SS cylinder and 1/4" diameter piston
  • Manufacturer defect warranty
More About this Item
Optos series piston pumps are designed to provide years of trouble-free service. Ensure the longevity of your pump by performing a few simple maintenance procedures. Each maintenance kit contains piston seals, check valve cartridges, retainer, filter, and tools.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Conformite Europeenne
Specifications
  • Compatible WithPumps 48531-62, -65, -73, -76, -84, and -87
  • DescriptionPiston Metering Pump Maintenance Kit for 316 SS Cylinder and 1/4" Dia Piston
  • WarrantyManufacturer defect