Edwards Tip Seal and Exhaust Service Kit for xDS35i Dry Scroll Vacuum Pumps
Representative image only.
Edwards Tip Seal and Exhaust Service Kit for xDS35i Dry Scroll Vacuum Pumps
Edwards | Mfr: A73001801| Item: 7930169
Repair kits help prolong the life of your vacuum pump
₹201,000.00INR / Each
₹201,000.00INR / Each
Product Description
Includes
1x spring 9.27 O/D x 19.5Lg, 2x tip seal XDS30, 3x washer plain Ss M4, 3x M4 X8 soc cap screw SS, 3x screw cap Hd Ss M4X12 A2-70, 6x screw M5 X20 Skt Caphd S/S, 1x O-ring FKM GP 19.6 ID x 2.4 sec, 6x washer,M5,S/coil, rect., 1x exhaust valve pad.
Key Features
  • Keep service parts handy to minimize downtime
  • Kit for 79301-33, -34, -35, -36, -37, -38, -39, and -40
  • Helps restore seal integrity and optimize vacuum performance
  • 1 year warranty
More About this Item
Genuine parts provide a perfect fit ensuring reliable performance.
1 Year Warranty
Specifications
  • Compatible WithxDS35i dry scroll vacuum pumps
  • TypeTip Seal and Exhaust Service Kit
  • DescriptionTip Seal and Exhaust Service Kit for xDS35i Dry Scroll Vacuum Pumps