Edwards NW25 to 28 mm Bore Tube Adapter
Representative image only.
Edwards NW25 to 28 mm Bore Tube Adapter
Edwards | Mfr: C10520201| Item: 7930057
Adapters provide quick and easy transitions to tubing or pipe thread
₹36,200.00INR / Each
₹36,200.00INR / Each
Product Description
Includes
KF25 NW25 C clamp and NW25 KF25 O-ring.
Key Features
  • NW25 flange connection
  • NW25 to 28 mm ID tubing
  • 1 year warranty
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithEM-series oil-sealed rotary vane vacuum pumps
  • ConnectionsNW25 to 28 mm OD hose barb
  • DescriptionNW25 to 28 mm Bore Tube Adapter