Edwards Inlet/Exhaust Filter NW25 for XDS and nXDS Dry Scroll Vacuum Pumps, 5 µm
Representative image only.
Edwards Inlet/Exhaust Filter NW25 for XDS and nXDS Dry Scroll Vacuum Pumps, 5 µm
Edwards | Mfr: A50597805| Item: 7930141
Place on inlet or outlet to protect your vacuum pump
₹222,000.00INR / Each
₹222,000.00INR / Each
Buy It Together
Current Item
Edwards Inlet/Exhaust Filter NW25 for XDS and nXDS Dry Scroll Vacuum Pumps, 5 µm
Edwards Inlet/Exhaust Filter NW25 for XDS and nXDS Dry Scroll Vacuum Pumps, 5 µm
7930141
₹222,000.00INR / Each
+
Edwards Inlet/exhaust filter spares - 5 micron element
7930154
₹41,700.00INR / Each
=
Total Price 263700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Supplied ready to use with a 5 µm filtering element
  • For XDS and nXDS pumps with NW25 port
  • 1 year warranty
More About this Item
Dual-purpose filter can be placed at the inlet or the outlet of your pump. Place between the pump inlet and your chamber outlet connection to prevent dust from entering into your system. Place it at the outlet between the vacuum pump and the exhaust line to trap out tip-seal dust.
1 Year Warranty
Specifications
  • Compatible WithXDS and nXDS dry scroll vacuum pumps with NW25 port
  • ConnectionsNW25
  • DescriptionInlet/Exhaust Filter NW25 for XDS and nXDS Dry Scroll Vacuum Pumps, 5 µm