Edwards Clean and Overhaul Kit for E2M28 Oil-Sealed Rotary Vane Vacuum Pumps
Representative image only.
Edwards Clean and Overhaul Kit for E2M28 Oil-Sealed Rotary Vane Vacuum Pumps
Edwards | Mfr: A37301131| Item: 7930068
Repair kits help prolong the life of your vacuum pump
₹124,000.00INR / Each
₹124,000.00INR / Each
Product Description
Includes
1x gasket E2M28, 2x O-ring NIT 44.5 ID x 3.0 sec, 1x O-ring NIT 29.5 ID x 3.0 sec, 2x O-ring FKM GP 15.6 ID x 2.4 sec, 1x O-ring FKM GP 13.6 ID x 2.4 sec, 1x O-ring NIT 35.5 ID x 3.0 sec, 1x O-ring NIT 79.5 ID x 3.0 sec, 1x O-ring NIT 107.63 x 2.62 dowty, 2x O-ring FKM GP 11.1 ID x 1.65 sec, 2x O-ring FKM GP 1.109 ID x .139 sec, 2x O-ring FKM GP 0.299 ID x .103 sec, 1x waved washer, 2x spring, 1x O-ring NIT 89.5 ID x 3.0 sec, 2x oil filter seal, 2x filter pad felt 1/4" thick, 2x ballast gauze E1M18, 1x label - Exhaust Warning, 1x filter assy E2M 18/28, 3x valve pad, 1x coupling element EM18/28/30, 1x spring, 7x O-ring NIT 6.6 ID x 2.4 sec, 1x spring, 2x seal FKM GP 25.35.6 UNILH, 1x dowty b/seal-size re.211 M8Scr, 1x distributor rubber FKM, 1x seal, 1x gasket, 1x oil box gasket E1M18, 1x dowty bonded seal MK3 2 BA, 1x felt pad, 1x O-ring FKM GP 14.1 ID x 1.6 sec, 1x O-ring FKM GP 69.5 ID x 3.0 sec, 1x O-ring FKM GP 194.5 ID x 3.0 sec.
Key Features
  • Keep service parts handy to minimize downtime
  • Service kit for 79300-39, 41, -42, -43, and -44
  • 1 year warranty
More About this Item
Genuine parts provide a perfect fit ensuring reliable performance.
The clean and overhaul kit contains all of the springs, gaskets, seals, and other components required for routine maintenance and whenever the pump is dismantled.
Fluoroelastomer clean and overhaul kit works with the following models:
E2M28 rotary vane pumps
1 Year Warranty
Specifications
  • Compatible WithE2M28 oil-sealed rotary vane vacuum pumps
  • TypeClean and Overhaul Kit
  • DescriptionClean and Overhaul Kit for E2M28 Oil-Sealed Rotary Vane Vacuum Pumps