Edwards Clean and Overhaul Kit for E2M0.7 and E2M1.5 Oil-Sealed Rotary Vane Pumps
Representative image only.
Edwards Clean and Overhaul Kit for E2M0.7 and E2M1.5 Oil-Sealed Rotary Vane Pumps
Edwards | Mfr: A37101131| Item: 7930051
Repair kits help prolong the life of your vacuum pump
₹93,700.00INR / Each
₹93,700.00INR / Each
Product Description
Includes
1x oil filter assembly stainless for E2M1.5, 1x valve pad E2M1.5 E2M18 pump, 1x O-ring FKM GP 6.6 ID x 2.4 sec, 1x O-ring NIT 25.5 ID x 3.0 sec, 1x spring E2M1.5, 1x seal FKM GP 16.30.7/6 Uni, 1x seal FKM GP 16.30.7 Pln, 2x O-ring NIT 52.3 ID x 5.7 sec, 1x seal FKM GP 16.30.7 Pos, 2x spring, 1x felt pad For E2M1.5, 1x ballast filter E2M1.5, 1x spring ballast EM, 2x spring Ez-2/5/8 Bo, 1x inlet strainer mesh, 1x compression spring, 1x O-ring NIT 12.6 ID x 2.4 sec, 3x O-ring NIT 21.6 ID x 2.4 sec, 3x drive member E2M1.5, 1x O-ring NIT 1.612 ID x.103 sec, 1x distributor valve, 1x O-ring NIT 11.1 ID x 1.6 sec, 1x O-ring 15M/M Idx 5M/M Sect Nit, 1x ballast valve, 4x O-ring FKM GP 0.299 ID x .103 sec, 1x oil box gasket E2M1.5, 1x oil drainplug 1/8" BSP, 1x O-ring NIT 1.109 ID x .139 sec, 1x O-ring NIT 15.6 ID x 2.4 sec, 1x O-ring NIT 124.5 ID x 3.0 sec, 1x oil drain plug RV, 1x sight glass gasket-E2M0.7/1.5.
Key Features
  • Keep service parts handy to minimize downtime
  • Service kit for 79300-27, -28, and -29
  • 1 year warranty
More About this Item
Genuine parts provide a perfect fit ensuring reliable performance.
The clean and overhaul kit contains all of the springs, gaskets, seals, and other components required for routine maintenance and whenever the pump is dismantled.
Fluoroelastomer clean and overhaul kit works with the following models:
E2M0.7 rotary vane pumps
E2M1.5 rotary vane pumps
1 Year Warranty
Specifications
  • Compatible WithE2M0.7 and E2M1.5 Oil-Sealed Rotary Vane Pumps
  • TypeClean and Overhaul Kit
  • DescriptionClean and Overhaul Kit for E2M0.7 and E2M1.5 Oil-Sealed Rotary Vane Pumps