Edwards C333-55-000 Vacuum isolation valve, NW 25 connections
Representative image only.
Edwards C333-55-000 Vacuum isolation valve, NW 25 connections
Edwards | Mfr: C333-55-000| Item: 7930260
₹77,700.00INR / Each
₹77,700.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
These manual vacuum valves are available as either a needle or isolation valve. The vacuum needle valve model is ideal for fine control or gas bleed. The vacuum isolation valve model is ideal for closing off sections of your vacuum system. An indicator displays open or closed position.
1 Year Warranty
Specifications
  • DescriptionVacuum isolation valve, NW 25 connections