Edwards C10516304 Vacuum Pump Fittings, clamp, NW 32 and 40
Representative image only.
Edwards C10516304 Vacuum Pump Fittings, clamp, NW 32 and 40
Edwards | Mfr: C10516304| Item: 3140044
₹7,200.00INR / Each
₹7,200.00INR / Each
Product Description
More About this Item
Achieve vacuum-tight seals with NW flange fittings by using a clamp with either a co-seal or a centering ring (order separately). Clamps are constructed of 316 SS, PTFE, VITON® fluoroelastomer, aluminum, and perbunen®. Temperature range is from 14 to 212°>F (–10 to 100°>C).
Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeVacuum Flange
  • Flange Size (Port 1)NW 40
  • DescriptionVacuum Pump Fittings, clamp, NW 32 and 40