Edwards C10514412 Vacuum Pump Fittings, aluminum cross, NW 25
Representative image only.
Edwards C10514412 Vacuum Pump Fittings, aluminum cross, NW 25
Edwards | Mfr: C10514412| Item: 3140062
₹30,600.00INR / Each
₹30,600.00INR / Each
Product Description
More About this Item
Choose from chemically resistant 316 stainless steel (SS) or economical aluminum.
A blank flange will seal an outlet not in use. Elbow fittings provide a 90°> bend. Tee and cross fittings let you connect two or three lines to your pump. Order clamps and either co-seals or centering rings below.
Specifications
  • Fitting MaterialAluminum
  • Connection TypeCross Vacuum Flange NW 25
  • Flange Size (Port 1)NW 25
  • Flange Size (Port 2)NW 25
  • Flange Size (Port 3)NW 25
  • Flange Size (Port 4)NW 25
  • DescriptionVacuum Pump Fittings, aluminum cross, NW 25