Edwards C105-14-411 Flange Tee Vacuum Pump Fitting, Aluminum, NW 25
Representative image only.
Edwards C105-14-411 Flange Tee Vacuum Pump Fitting, Aluminum, NW 25
Edwards | Mfr: C105-14-411| Item: 3140032
₹22,800.00INR / Each
₹22,800.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Choose from chemically resistant 316 stainless steel (SS) or economical aluminum.
A blank flange will seal an outlet not in use. Elbow fittings provide a 90°> bend. Tee and cross fittings let you connect two or three lines to your pump. Order clamps and either co-seals or centering rings below.
1 Year Warranty
Specifications
  • Fitting MaterialAluminum
  • Connection TypeVacuum Flange
  • Flange Size (Port 1)NW 25
  • Flange Size (Port 2)NW 25
  • Flange Size (Port 3)NW 25
  • DescriptionFlange Tee Vacuum Pump Fitting, Aluminum, NW 25