Edwards C105-14-304 Flanged Fitting Vacuum Clamp, for NW 20 or 25
Representative image only.
Edwards C105-14-304 Flanged Fitting Vacuum Clamp, for NW 20 or 25
Edwards | Mfr: C105-14-304| Item: 3140042
₹7,800.00INR / Each
₹7,800.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Achieve vacuum-tight seals with NW flange fittings by using a clamp with either a co-seal or a centering ring (order separately). Clamps are constructed of 316 SS, PTFE, VITON® fluoroelastomer, aluminum, and perbunen®. Temperature range is from 14 to 212°>F (–10 to 100°>C).
1 Year Warranty
Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeVacuum Flange
  • Flange Size (Port 1)NW 25
  • DescriptionFlanged Fitting Vacuum Clamp, for NW 20 or 25