Edwards C105-12-393 Centering ring, NW 10/16 - Nitrile (Vacuum Pump Accessories)
Representative image only.
Edwards C105-12-393 Centering ring, NW 10/16 - Nitrile (Vacuum Pump Accessories)
Edwards | Mfr: C105-12-393| Item: 3140270
Create a tight vacuum seal with these centering rings
₹3,400.00INR / Each
₹3,400.00INR / Each
Product Description
More About this Item
Polymer support and nitrile O-rings withstand up to 10–7 Torr (mm Hg). The temperature range of these centering rings is 14 to 212°F (–10 to 100°C).
The centering ring has an elastomer O-ring creating the seal, and inside of this there is a plastic support ring.
Specifications
  • Fitting MaterialNitrile
  • Connection TypeVacuum Flange
  • Flange Size (Port 1)NW 16
  • DescriptionCentering ring, NW 10/16 - Nitrile (Vacuum Pump Accessories)