Edwards C105-04-104 Adapter, SMOOTH, NW 16 to 1/2" hose barb, 316L SS

Representative image only.

Edwards C105-04-104 Adapter, SMOOTH, NW 16 to 1/2" hose barb, 316L SS

Edwards | Mfr: C105-04-104| Item: 3140175

Use with a clamp and co-seal or centering ring

₹13,000.00INR / Each

₹13,000.00INR / Each

Buy It Together

Current Item

Edwards C105-04-104 Adapter, SMOOTH, NW 16 to 1/2" hose barb, 316L SS
Edwards C105-04-104 Adapter, SMOOTH, NW 16 to 1/2" hose barb, 316L SS
3140175

₹13,000.00INR / Each
+ =
Total Price 19100.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring.

Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeVacuum Flange x Barb
  • Barb Size / Tubing Id (Port 2) (in)1/2
  • Flange Size (Port 1)NW 16
  • DescriptionAdapter, SMOOTH, NW 16 to 1/2" hose barb, 316L SS
Reviews
Q&A