Edwards C105-01-103 Vacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"NPT(M)

Representative image only.

Edwards C105-01-103 Vacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"NPT(M)

Edwards | Mfr: C105-01-103| Item: 3140103

Use with a clamp and co-seal or centering ring

₹55,100.00INR / Each

₹55,100.00INR / Each

Buy It Together

Current Item

Edwards C105-01-103 Vacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"NPT(M)
Edwards C105-01-103 Vacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"NPT(M)
3140103

₹55,100.00INR / Each
+ =
Total Price 61200.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring.

Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeVacuum Flange x NPT(M)
  • Thread Size NPT(M) (Port 2) (InchShort)1/4
  • Flange Size (Port 1)NW 16
  • DescriptionVacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"NPT(M)
Reviews
Q&A