Edwards C100-07-173 Centering Ring, 63 ISO-K - Nitrile (Vacuum Pump Accessories)
Representative image only.
Edwards C100-07-173 Centering Ring, 63 ISO-K - Nitrile (Vacuum Pump Accessories)
Edwards | Mfr: C100-07-173| Item: 3140182
Create a tight vacuum seal with these centering rings
₹13,000.00INR / Each
₹13,000.00INR / Each
Product Description
More About this Item
Polymer support and nitrile O-rings withstand up to 10–7 Torr (mm Hg). The temperature range of these centering rings is 14 to 212°F (–10 to 100°C).
The centering ring has an elastomer O-ring creating the seal, and inside of this there is a plastic support ring.
Specifications
  • Fitting MaterialNitrile
  • Connection TypeVacuum Flange
  • Flange Size (Port 1)63 ISO-K
  • DescriptionCentering Ring, 63 ISO-K - Nitrile (Vacuum Pump Accessories)