Edwards C100-07-156 Vacuum Pump Fittings, Clamp, 63 and 100 ISO-K
Representative image only.
Edwards C100-07-156 Vacuum Pump Fittings, Clamp, 63 and 100 ISO-K
Edwards | Mfr: C100-07-156| Item: 3140046
₹3,700.00INR / Each
₹3,700.00INR / Each
Product Description
Key Features
  • Constructed of 316 SS, PTFE, Viton® fluoroelastomer, aluminum, and perbunen
More About this Item
Achieve vacuum-tight seals with NW flange fittings by using a clamp with either a co-seal or a centering ring (order separately). Clamps are constructed of 316 SS, PTFE, VITON® fluoroelastomer, aluminum, and perbunen®. Temperature range is from 14 to 212°>F (–10 to 100°>C).
Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeVacuum Flange
  • Flange Size (Port 1)100 ISO-K
  • DescriptionVacuum Pump Fittings, clamp, 63 and 100 ISO-K