Edwards Bearing Replacement Kit for nxDS Dry Scroll Vacuum Pumps
Representative image only.
Edwards Bearing Replacement Kit for nxDS Dry Scroll Vacuum Pumps
Edwards | Mfr: A73501802| Item: 7930148
Repair kits help prolong the life of your vacuum pump
₹506,000.00INR / Each
₹506,000.00INR / Each
Product Description
Includes
4x thick Shim M.S. 1.4 mm, 4x thick Shim M.S. 1.1 mm, 8x thin Shim St.Stl 0.075 mm, 8x thin Shim St.Stl 0.050 mm, 1x deep groove bearing 6206, 1x NxDS rear bearing-carrier assy, 4x push rivet blk nylon dia 5 x 11, 2x O-ring FKM GP 1.109 ID x .139 sec, 1x OS bearing Mkii, 1x gasket, rear bearing cap, 4x O-ring FKM GP 19.6 ID x 2.4 sec, 1x O-ring FKM GP 0.299 ID x .103 sec, 1x O-ring FKM GP 7.5 ID x 1.5 sec
Key Features
  • Keep service parts handy to minimize downtime
  • Kit for 79301-19 through -31 pumps
  • Helps optimize vacuum performance and reduce noise and vibration
  • 1 year warranty
More About this Item
Genuine parts provide a perfect fit ensuring reliable performance.
1 Year Warranty
Specifications
  • Compatible WithnxDS dry scroll vacuum pumps
  • TypeBearing Replacement Kit
  • DescriptionBearing Replacement Kit for nxDS Dry Scroll Vacuum Pumps