Edwards B271-58-448 Vacuum Pump Fittings, co-seal, NW 20 and 25
Representative image only.
Edwards B271-58-448 Vacuum Pump Fittings, co-seal, NW 20 and 25
Edwards | Mfr: B271-58-448| Item: 3140262
Eliminate crevices and trapped volumes in your line
₹8,900.00INR / Each
₹8,900.00INR / Each
Product Description
Key Features
  • Polymer/nitrile construction
  • 1 year warranty
More About this Item
Co-seals eliminate crevices and trapped volumes in your line. Polymer support and nitrile O-rings withstand up to 10–7 torr (mm Hg). Temperature range is from 14 to 176°F
(–10 to 80°C).
1 Year Warranty
Specifications
  • Fitting MaterialPolymer
  • Connection TypeVacuum Flange
  • Flange Size (Port 1)NW 20/25
  • DescriptionVacuum Pump Fittings, co-seal, NW 20 and 25