Edwards A441-10-000 Vacuum Pump Accessories, Inlet Condensate Trap, NW 25
Representative image only.
Edwards A441-10-000 Vacuum Pump Accessories, Inlet Condensate Trap, NW 25
Edwards | Mfr: A441-10-000| Item: 7930200
₹308,000.00INR / Each
₹308,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Inlet condensate trap protects pumps from harmful vapors. Wetted parts are glass, aluminum, and VITON. NW 25 ports. Capacity is 1.84 liters. Measures 8-1/4"L x 6-3/4" dia.
1 Year Warranty
Specifications
  • DescriptionVacuum Pump Accessories, Inlet Condensate Trap, NW 25