Eco Sensors C-30ZX Ozone Monitor with Alarm

Representative image only.

Eco Sensors C-30ZX Ozone Monitor with Alarm

Eco Sensors | Mfr: C-30ZX| Item: 8631610

Fixed system alerts workers to hazardous ozone levels

₹196,000.00INR / Each

₹196,000.00INR / Each

Buy It Together

Current Item

Eco Sensors C-30ZX Ozone Monitor with Alarm
Eco Sensors C-30ZX Ozone Monitor with Alarm
8631610

₹196,000.00INR / Each
+
Eco Sensors OG-3 Ozone Calibrator; 0.1 ppm
8631661

₹179,000.00INR / Each
+ =
Total Price 375000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Easy-to-read, multicolor graphical display
  • HMOS sensor for superior sensitivity
  • 1 year warranty

More About this Item

LED bargraph changes color as ozone levels increase: green indicates a safe level, yellow is caution level, and red is the danger level. Features a heated metal oxide semiconductor (HMOS) sensor for excellent sensitivity and quick response time. Also features an audio alarm and relay output that are triggered at 0.1 ppm (danger level) and a 0 to 3 VAC recorder output. Order optional calibrator to verify that your ozone detectors are conforming to their origianl calibration.

1 Year Warranty
Specifications
  • Gases MeasuredOzone
  • AlarmAudible
  • Height (in)3 1/4
  • Width (in)2 3/8
  • Depth (in)1 3/8
  • DescriptionOzone Monitor with Alarm
  • Warranty1 year
Reviews
Q&A