Eco Sensors C-21 Datalogging VOC Detector with Alarm

Representative image only.

Eco Sensors C-21 Datalogging VOC Detector with Alarm

Eco Sensors | Mfr: C-21| Item: 8631600

Prevent health hazards by detecting leaks and poor air quality early

Call For Price

Call For Price

Buy It Together

Current Item

Eco Sensors C-21 Datalogging VOC Detector with Alarm
Eco Sensors C-21 Datalogging VOC Detector with Alarm
8631600

Call For Price
+
Eco Sensors OG-3 Ozone Calibrator; 0.1 ppm
8631661

₹179,000.00INR / Each
=
Total Price 179000.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Detects acetone, methylene chloride, naphtha, trichloroethylene, and more
  • Ideal for laboratories, paint/print shops, and woodworking facilities
  • 1 year warranty

More About this Item

Use this economical VOC sensor wherever solvents are present—ideal for paint and print shops, wood working, and laboratories. The sensor is highly sensitive to VOCs and solvent vapors such as methylene chloride, acetone, naphtha, perchloroethylene, and trichloroethylene. The LED bar graph displays the hazard level of gas or vapor—green indicates normal, yellow for caution, and red for danger. An audible alarm also alerts you to dangerous levels. Responds in one minute. A rechargeable NiCd battery lets you take the detector anywhere—trace leaks to their source. Just plug the detector in and you're ready to go—nothing to install. Measures 23/8''W x 31/2''H x 13/8''D.

1 Year Warranty
Specifications
  • Gases MeasuredVolatile organic compounds
  • Display TypeLED bar graph
  • Battery Type1 nickel-cadmium rechargeable (included)
  • Height (in)3 1/2
  • Width (in)2 3/8
  • Depth (in)1 3/8
  • DescriptionDatalogging VOC Detector with Alarm
  • Warranty1 year
Reviews
Q&A