Eagle Stainless Storage Tank with Clip-Down Cover, Stainless Steel, 5.2 Gal
Representative image only.
Eagle Stainless Storage Tank with Clip-Down Cover, Stainless Steel, 5.2 Gal
Eagle Stainless | Mfr: CTH-30 T-316L| Item: 0750520
Provides air/water-tight seal
₹299,000.00INR / Each
₹299,000.00INR / Each
Buy It Together
Current Item
Eagle Stainless Storage Tank with Clip-Down Cover, Stainless Steel, 5.2 Gal
Eagle Stainless Storage Tank with Clip-Down Cover, Stainless Steel, 5.2 Gal
0750520
₹299,000.00INR / Each
+ =
Total Price 378000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Seamless sanitary interior makes clean up easy
  • 3 month warranty
More About this Item
These 316L stainless steel tanks feature clip-down lid, handles, and platinum cured silicone gasket. Clean up is made easy with seamless sanitary interior and exterior with radiused bottom-corners.
3 Month Warranty
Specifications
  • MaterialStainless Steel
  • Volume (Gal)5.2
  • Volume (Liters)20
  • Height (in)11 51/64
  • Height (cm)30
  • DescriptionStainless Steel Storage Tank with Clip-down Cover, 5.2 gal