Eagle Stainless Storage Tank with Clip-Down Cover, Stainless Steel, 26.4 Gal
Representative image only.
Eagle Stainless Storage Tank with Clip-Down Cover, Stainless Steel, 26.4 Gal
Eagle Stainless | Mfr: CTH-47H T-316L| Item: 0750510
Provides air/water-tight seal
₹820,000.00INR / Each
₹820,000.00INR / Each
Buy It Together
Current Item
Eagle Stainless Storage Tank with Clip-Down Cover, Stainless Steel, 26.4 Gal
Eagle Stainless Storage Tank with Clip-Down Cover, Stainless Steel, 26.4 Gal
0750510
₹820,000.00INR / Each
+ =
Total Price 994000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Seamless sanitary interior makes clean up easy
  • 3 month warranty
More About this Item
These 316L stainless steel tanks feature clip-down lid, handles, and platinum cured silicone gasket. Clean up is made easy with seamless sanitary interior and exterior with radiused bottom-corners.
3 Month Warranty
Specifications
  • MaterialStainless Steel
  • Volume (Gal)26.4
  • Volume (Liters)100
  • Height (in)23 19/32
  • Height (cm)60
  • DescriptionStainless Steel Storage Tank with Clip-down Cover, 26.4 gal