Eagle M24CRA Modular Quik-Assembly Acid and Corrosive Safety Cabinet, 4 Shelves, 24 Gal
Representative image only.
Eagle M24CRA Modular Quik-Assembly Acid and Corrosive Safety Cabinet, 4 Shelves, 24 Gal
Eagle | Mfr: M24CRA| Item: 8177328
Assembles in minutes with no tools required
₹261,000.00INR / Each
₹261,000.00INR / Each
Product Description
Key Features
  • Stores corrosive liquids safety
  • Easy to store
  • Sealed internal containment sump
  • 1 year warranty
More About this Item
Modular all-HDPE cabinets are designed to safely store corrosive or aggressive liquids in labs and industrial locations. Assembly is simple, and when not in use, cabinets can be broken down in minutes without tools and easily stored. The stand-alone cabinets can be assembled with the option of the lockable door opening to the right or left.
Can be linked and stacked with additional 12 Gallon units to create additional vertical or horizontal design. Features a 2.1 gallon sealed internal containment sump with dimension of 14.5" W x 18.5" D x 1.75" H.
1 Year Warranty
Specifications
  • MaterialPE
  • Capacity (Gal)24
  • Number of Shelves4
  • Width (in)18
  • Height (in)72
  • Depth (InchShort)22
  • ColorBlue
  • Self-ClosingNo
  • Shipping Weight (lbs)90
  • DescriptionModular Quik-Assembly Safety Cabinet, 24 Gal., 2 Doors, 4 Shelves - Blue