Eagle 1924X Flammable Liquid Safety Cabinet, Space Saver, Self Closing, 1 Door, 12 Gal

Representative image only.

Eagle 1924X Flammable Liquid Safety Cabinet, Space Saver, Self Closing, 1 Door, 12 Gal

Eagle | Mfr: 1924X| Item: 0941822

Adjustable steel shelves hold up to 350 lb

₹272,000.00INR / Each

₹272,000.00INR / Each
Product Description

Key Features

  • Four adjustable galvanized leveling legs
  • Trilingual warning labels
  • Raised leakproof sill
  • Meeta NFPA Code 30 and OSHA requirements

More About this Item

Flammable cabinets are constructed of a high gloss powder coated finish, double-walled, 18-gauge steel with 1½" (3.8 cm) insulating air barrier — ideal for safely storing flammable liquid containers up to 5 gallons in size. Cabinets feature dual vents, each have 2" (5.1 cm) threaded fittings and a fire baffle and cap, grounding attachments, red trilingual warning, 2" (5.1 cm) raised, leakproof door sill, 3-point latch system, and flush mounted door handle with key lock. A concealed self-close mechanism provides obstruction-free access to top shelf space.

Includes adjustable galvanized steel shelves which direct spills to back and bottom of leakproof sump and hold 350 lbs (158.8 kg) Uniform Distributed Load (UDL). Hazard label: FLAMMABLE-KEEP FIRE AWAY.

Fetrures self-closing doors feature a fusible link that is actuated at 160°F/71°C.

FM Approved
Specifications
  • Material18-gauge steel
  • Capacity (Liters)45.4
  • Capacity (Gal)12
  • Number of Shelves1
  • Width (in)23
  • Height (in)35
  • Depth (InchShort)18
  • Width (cm)58.4
  • Height (cm)88.9
  • Depth (cm)45.7
  • ColorYellow
  • Self-ClosingYes
  • ComplianceOSHA, NFPA Code 30, UFC 79
  • Shipping Weight (lbs)108
  • DescriptionFlammable Safety Cabinet, Self Closing, 12 gallon
Reviews
Q&A