Eagle 1650 Qverpack Salvage Plastic Drum Barrel, Screw-On Lid, 20 Gal

Representative image only.

Eagle 1650 Qverpack Salvage Plastic Drum Barrel, Screw-On Lid, 20 Gal

Eagle | Mfr: 1650| Item: 8611004

For secondary storage, containment, transport, or clean-up of 55-gallon drums or smaller containers

₹16,600.00INR / Each

₹16,600.00INR / Each
Product Description

Key Features

  • Passed a 3-PSI pressure test.
  • UN HM-181 approved

More About this Item

The salvage drums are constructed of yellow blow-molded high-density polyethylene (HDPE) with UV inhibitors, are durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration.
Drums are available with a metal band with bolt closure or screw top lids.

Specifications
  • Capacity (Liters)75.7
  • Capacity (Gal)20
  • Height (in)21 1/2
  • Height (cm)54.6
  • ComplianceDOT 49 CFR-173.3(c); 3psi pressure test for Salvage Drum Compliance
  • Shipping Weight (lbs)12.8
  • DescriptionOverpack Salvage/Secondary Drum, 20 gallon
Reviews
Q&A