Eagle 1613 Drum Bogie Dolly
Representative image only.
Eagle 1613 Drum Bogie Dolly
Eagle | Mfr: 1613| Item: 8639400
Carrier protects your plant floor from messy drum leaks
₹72,000.00INR / Each
₹72,000.00INR / Each
Product Description
Key Features
  • Recessed spill well holds up to 12 gallons
  • Easily transport heavy drums around the plant
  • Fits 30, 55 and 95 Gal drums
  • 1 year warranty
More About this Item
The durable drum bogie is constructed of high density polyethylene (HDPE) and has a load capacity of 1,250 lbs. This mobile dispensing unit protects against incidental spills and leaks for 30, 55, and 95 gallon drums. It resists corrosion, has five swivel casters for maximum stability, load capacity, and maneuverability. The integrated push-pull handle locks in vertical position.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • Height (in)42
  • Height (cm)106.7
  • Shipping Weight (lbs)30
  • DescriptionYellow PE Mobile Drum Spill Bogie/Cart