E Instruments E85AACSF13 Optional 12" Probe with 10-ft dual hose for Portable Indoor Air Quality Monitor
Representative image only.
E Instruments E85AACSF13 Optional 12" Probe with 10-ft dual hose for Portable Indoor Air Quality Monitor
E Instruments | Mfr: E85AACSF13| Item: 8197411
Optional probe to perform more complete analyses in the field
₹82,800.00INR / Each
₹82,800.00INR / Each
Product Description
Key Features
  • Ideal for labs, cleanrooms, schools, and much more
  • 1 year warranty
More About this Item
Optional remote probe with 10-ft (3-m) hose for use in hard-to-reach areas. Probe is good for temperatures up to 1470°F (800°C) and includes a built-in thermocouple for temperature measurement.
Conformite Europeenne1 Year Warranty
Specifications
  • Compatible With81974-00, 81974-01, 81974-02, 81974-03
  • DescriptionOptional 12" Probe with 10-ft dual hose for Portable Indoor Air Quality Monitor