Dynalon Disposable Stirring Rod/Spatulas, PS; 1000/Cs
Representative image only.
Dynalon Disposable Stirring Rod/Spatulas, PS; 1000/Cs
Dynalon | Mfr: 375985| Item: 0634335
Compatible with cuvettes and other micro-volume containers
₹10,400.00INR / Case of 1000
₹10,400.00INR / Case of 1000
Product Description
More About this Item
Disposable polystyrene cuvette stirring spatulas measure 120 mm long by 3 mm in diameter. Packed 500 stirrers in a polyethylene bag; 2 bags (1000 stirrers) per case.
Specifications
  • Overall Length (in)4 3/4
  • Overall Length (cm)12
  • MaterialPolystyrene
  • StyleCuvettes and other micro-volume containers
  • DescriptionDisposable Stirring Rod/Spatulas, PS; 1000/Cs