Dynalon 797303-6300 M-Stand for 30L Bio-bin Waste Disposal Containers

Representative image only.

Dynalon 797303-6300 M-Stand for 30L Bio-bin Waste Disposal Containers

Dynalon | Mfr: 797303-6300| Item: 8140212

₹66,400.00INR / Each

₹66,400.00INR / Each
Product Description

Key Features

  • Holds 30 L Bio-bin

More About this Item

These support stands safely support Bio-bin® waste disposal containers.

M stand holds the 30 L Bio-bin. Stand is supplied with four (4) non-slip feet. Stainless steel.

Specifications
  • MaterialStainless Steel
  • Capacity (Liters)30
  • DescriptionM-Stand for 30L Bio-bin (Material: Stainless Steel)
Reviews
Q&A