Dynalon 797303-6200 Z-Stand for 6L Bio-bin Waste Disposal Containers

Representative image only.

Dynalon 797303-6200 Z-Stand for 6L Bio-bin Waste Disposal Containers

Dynalon | Mfr: 797303-6200| Item: 8140211

₹19,100.00INR / Each

₹19,100.00INR / Each
Product Description

Key Features

  • Holds 6 L Bio-bin upright

More About this Item

These support stands safely support Bio-bin® waste disposal containers.

Z stand holds the 6 L Bio-bin upright, either for bench or floor use and allows for easy removal of used bins. Stand is supplied with four (4) non-slip feet. Stainless steel.

Specifications
  • Capacity (Liters)6
  • MaterialStainless Steel
  • DescriptionZ-Stand for 6L Bio-bin (Stands for Bio-bin Waste Disposal Containers)
Reviews
Q&A