Dynalon 797303-6100 U-Stand for 6L Bio-bin Waste Disposal Containers

Representative image only.

Dynalon 797303-6100 U-Stand for 6L Bio-bin Waste Disposal Containers

Dynalon | Mfr: 797303-6100| Item: 8140210

₹19,100.00INR / Each

₹19,100.00INR / Each
Product Description

Key Features

  • Holds 6 L Bio-bin at 30° angle

More About this Item

These support stands safely support Bio-bin® waste disposal containers.

U stand holds the 6 L Bio-bin at a 30° angle, making it ideal for use in a fume cabinet, biological safety cabinet, other workstation or bench. It angles the bin so that laboratory pipettes can be disposed easily. Stand is supplied with four (4) non-slip feet. Stainless steel.

Specifications
  • Capacity (Liters)6
  • MaterialStainless Steel
  • DescriptionU-Stand for 6L Bio-bin (Stands for Bio-bin Waste Disposal Containers)
Reviews
Q&A