Dynalon 797303-0002 Bio-bin Waste Disposal Container, 2.5L, Case of 50

Representative image only.

Dynalon 797303-0002 Bio-bin Waste Disposal Container, 2.5L, Case of 50

Dynalon | Mfr: 797303-0002| Item: 8140207

Great alternative to plastic waste collection and disposal containers

₹90,900.00INR / Case of 50

₹90,900.00INR / Case of 50
Product Description

Includes

2" x 2" "biohazard" waste labels.

Key Features

  • Space-efficient, lightweight fold-flat design
  • Waterproof construction — inside and outside — with heat-sealed joints
  • Built-in super absorbent Bio-matt™ liner soaks up excess liquid

More About this Item

These disposable plastic-coated paper containers are perfect for laboratory waste that requires special heat treatment such as autoclaving or incineration before disposal. The built-in lid has both a temporary and permanent seal and features a handle for easy transport when full. Ideal for use in academic, clinical, government, and industrial laboratories for a wide range of non-sharp biohazard or non-hazardous waste. Autoclavable

This new bench top size is ideal for disposal of non-sharps biomedical and laboratory waste, up to 9" (228mm) in length such as microbiological inoculating loops, disposable, single use instruments, transfer pipettes, swabs. The longer base allows waste to lie flat on the bottom reducing wastage. Built-in Bio-matt absorbs approx 1/2 liter of liquid. Items such as microbiological loops, pipettes, tips, test tubes, swabs and vials are ideal for disposal by this method.

Specifications
  • Capacity (Liters)2.5
  • Length (in)9
  • Width (in)4.3125
  • Length (cm)22.86
  • Width (cm)10.95375
  • DescriptionBio-bin Waste Disposal Container, 2.5L, Case of 50
Reviews
Q&A