DWK Life Sciences (Wheaton) W348923-C Mini Bottle Benchtop Roller Culture System, 230V Continental Europe

Representative image only.

DWK Life Sciences (Wheaton) W348923-C Mini Bottle Benchtop Roller Culture System, 230V Continental Europe

DWK Life Sciences (Wheaton) | Mfr: W348923-C| Item: 2930058

Use with standard incubators and cold rooms – small footprint is perfect for labs with limited space!

₹868,000.00INR / Each

₹868,000.00INR / Each

Buy It Together

Current Item

DWK Life Sciences (Wheaton) W348923-C Mini Bottle Benchtop Roller Culture System, 230V Continental Europe
DWK Life Sciences (Wheaton) W348923-C Mini Bottle Benchtop Roller Culture System, 230V Continental Europe
2930058

₹868,000.00INR / Each
+ =
Total Price 1166000.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Self-lubricated bearings eliminate routine maintenance issues
  • Attach up to two additional deck kits for versatility depending on your applications
  • 1 year warranty

More About this Item

Obtain better distribution of media and uniform gassing with these benchtop rollers, designed to continuously rotate cylindrical bottles at optimum speeds. Transistorized solid state circuitry allows exact speed control of the bottles and is designed to compensate against sudden or prolonged voltage changes. Roller shafts are mounted in self-lubricated nylon bearings, which eliminate many maintenance problems. Order bottles separately.

Mini bottle benchtop roller culture systems are designed for small-scale mixing and agitation using four 30 mL or larger serum bottles, 100-125 mL media bottles, or 38 x 200 mm culture tubes. These will accommodate bottles 38 to 60mm in diameter, up to 240mm long, and provides speeds of 3 to 45 rpm. Order up to two additional deck kits to increase your rolling capacity.

Conformite Europeenne1 Year WarrantyCanadian Standards AssociationUL Listed.
Specifications
  • Number of Bottles Accepted4
  • Min Bottle Diameter (mm)38
  • Max Bottle Diameter (mm)240
  • Max Bottle Length (mm)240
  • Bottle Speed (rpm)3 to 45 (38 mm bottle), 2 to 30 (60 mm bottle)
  • Width (in)12 1/4
  • Height (in)7 1/8
  • Length (in)12 3/4
  • Width (cm)31.1
  • Height (cm)18.1
  • Length (cm)32.4
  • Plug TypeEU
  • Power (VAC)230
  • Power (watts)14
  • Power (Hz)50/60
  • DescriptionMini Bottle Benchtop Roller Culture System, 230V Continental Europe
  • Warranty1 year
Reviews
Q&A