DWK Life Sciences (Wheaton) W225714 Vial Decapper, 13 mm, with Rechargeable Battery
Representative image only.
DWK Life Sciences (Wheaton) W225714 Vial Decapper, 13 mm, with Rechargeable Battery
DWK Life Sciences (Wheaton) | Mfr: W225814| Item: 9880006
Reduce crimping and decapping time
Call For Price
Call For Price
Buy It Together
Current Item
DWK Life Sciences (Wheaton) W225714 Vial Decapper, 13 mm, with Rechargeable Battery
DWK Life Sciences (Wheaton) W225714 Vial Decapper, 13 mm, with Rechargeable Battery
9880006
Call For Price
+
DWK Life Sciences (Wheaton) W225701 Crimping Tool Stand
9880024
₹75,500.00INR / Each
=
Total Price 75500.00 ADD 2 ITEMS TO CART
Product Description
Includes
Rechargeable battery and universal power supply.
Key Features
  • Ergonomically designed to reduce strain
  • Use with flip-off caps
  • 1 year warranty
More About this Item
Quickly and easily crimp and decap laboratory vials and seals with aluminum or two-part aluminum/steel caps. These battery-powered crimping tools come with a powerful motor with a fast cycle. Long lasting battery provides up to 800 crimps per charge and even lets you operate while charging. The electronically adjustable cycle is accurate and reproducible and can be adjusted with control pad on top of the unit. Use with optional tool stand to reduce hand fatigue.
1 Year Warranty
Specifications
  • Cap Diameter (mm)13
  • DescriptionBattery-powered decapper