DWK Life Sciences (Wheaton) 237071 Replacement Wick for Alcohol Burner, 6" L x 1/4" Dia
Representative image only.
DWK Life Sciences (Wheaton) 237071 Replacement Wick for Alcohol Burner, 6" L x 1/4" Dia
DWK Life Sciences (Wheaton) | Mfr: 237071| Item: 4858586
Ideal for drying slides
₹740.00INR / Each
₹740.00INR / Each
Product Description
Includes
Ground glass cap and one wick.
Key Features
  • Can use with isopropyl or denatured ethyl alcohol
  • Low-heat flame
  • Replacement wick for Alcohol Burner 48585-84
More About this Item
Manufactured from soda-lime glass, this burner is designed for use with isopropyl or denatured ethyl alcohol. The low-heat flame is well suited for microscopy purposes.
Specifications
  • Length (in)72
  • DescriptionReplacement Wick for Alcohol Burner, 6" L x 1/4" Dia