DWK Life Sciences (Wheaton) 224177-01 Open Top Aluminum Seals, Natural, 13mm, 1000/CS

Representative image only.

DWK Life Sciences (Wheaton) 224177-01 Open Top Aluminum Seals, Natural, 13mm, 1000/CS

DWK Life Sciences (Wheaton) | Mfr: 224177-01| Item: 0891826

Unlined aluminum vial seals are available in a wide range of styles and sizes

₹29,100.00INR / Case of 1000

₹29,100.00INR / Case of 1000
Product Description

Key Features

  • Constructed of durable aluminum

More About this Item

Open top seals feature an opening to allow for exposure of the vial's stopper or septa. Center disc tear-out seals feature a tear away center disc which allows for this exposure.

Specifications
  • Cap colorNatural
  • Height (in)33/64
  • Height (mm)13
  • DescriptionOpen Top Aluminum Seals, Natural, 13mm, 1000/CS
Reviews
Q&A