DWK Life Sciences (Kimble) Petri Dish Cover, 100 x 10 mm, 12/Cs
Representative image only.
DWK Life Sciences (Kimble) Petri Dish Cover, 100 x 10 mm, 12/Cs
DWK Life Sciences (Kimble) | Mfr: 23062-10010| Item: 1595834
Shallow glass dishes designed to meet Federal Specification NNN-D-1478
Call For Price
Call For Price
Product Description
Key Features
  • -Autoclavable
  • -Colored ceramic enamel marking spot on covers and bottom
  • -Includes reference point to locate serial dilution starting points on bottom
  • -Manufactured from 33 expansion, low extractable borosilicate glass confirming to USP Type I and ASTM E438,
  • Type I Class A requirements
More About this Item
KIMAX® dishes remain clear after repeated use in wet or dry sterilization cycles.
Reinforced beaded edges resist mechanical breakage
Combination of different colors and wording permits quick and easy identification of the parts
Specifications
  • DescriptionPetri Dish Cover, 100 x 10 mm, 12/Cs