DWK Life Sciences (Kimble) Mojonnier Glass Extraction Flask, 25 mL; 1/Pk
Representative image only.
DWK Life Sciences (Kimble) Mojonnier Glass Extraction Flask, 25 mL; 1/Pk
DWK Life Sciences (Kimble) | Mfr: 617600-0025| Item: 3459065
₹34,100.00INR / Each
₹34,100.00INR / Each
Product Description
More About this Item
Use this flask to measure fat content in dairy and other food products. Extraction chamber has a diameter of 35 mm; top opening accepts a #0 stopper.
Specifications
  • MaterialGlass
  • Capacity (mL)25
  • DescriptionMojonnier Glass Extraction Flask, 25 mL; 1/Pk