DWK Life Sciences (Kimble) KimCote® Class A Volumetric Flask with Glass Stopper; 2000 mL

Representative image only.

DWK Life Sciences (Kimble) KimCote® Class A Volumetric Flask with Glass Stopper; 2000 mL

DWK Life Sciences (Kimble) | Mfr: KC28014-2000| Item: 3452439

KimCote™ plastic coating for added safety

₹151,000.00INR / Case of 2

for an additional
₹27,300.00INR / Each
for an additional
₹39,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹151,000.00INR / Case of 2

Buy It Together

Current Item

DWK Life Sciences (Kimble) KimCote® Class A Volumetric Flask with Glass Stopper; 2000 mL
DWK Life Sciences (Kimble) KimCote® Class A Volumetric Flask with Glass Stopper; 2000 mL
3452439

₹151,000.00INR / Case of 2
+
InnoCal NIST-Traceable Calibration; Hydrometer, 1-point
1710700

₹27,300.00INR / Each
=
Total Price 178300.00 ADD 2 ITEMS TO CART
Product Description

Includes

volumetric flask and stopper.

Key Features

  • Reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Volumetric flasks are constructed of low extractable borosilicate glass
  • Volumetric flask features 2000 mL (64 oz) capacity with ±0.50 mL tolerance
  • Features 15.75" (40 cm) height
  • Includes no.27 glass stopper

More About this Item

Volumetric plastic-coated flasks feature white ceramic enamel marking spots and glass stoppers. Calibrated "To Contain", flasks include a single graduation ring. Flasks are designed to meet ASTM Spec E288, Class A unserialized requirements. Autoclavable to 120°C (250°F).

Specifications
  • MaterialGlass
  • Capacity (mL)2000
  • Tolerance (mL)±0.5
  • Cap TypePennyhead stopper
  • Stopper Number Accepted27
  • Overall Height (cm)40
  • DescriptionVolumetric Flask with Glass Stopper; 2000 mL
Reviews
Q&A