DWK Life Sciences (Kimble) Kimax Soxhlet Round Bottom Flask; 500 mL; 1/Pk
Representative image only.
DWK Life Sciences (Kimble) Kimax Soxhlet Round Bottom Flask; 500 mL; 1/Pk
DWK Life Sciences (Kimble) | Mfr: 601000-0624| Item: 9903766
Choose an extraction flask by volume and bottom shape
₹13,900.00INR / Each
₹13,900.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Kimble) Kimax Soxhlet Round Bottom Flask; 500 mL; 1/Pk
DWK Life Sciences (Kimble) Kimax Soxhlet Round Bottom Flask; 500 mL; 1/Pk
9903766
₹13,900.00INR / Each
+ =
Total Price 96900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Flat bottom flasks sit easily on flat surfaces
  • 1 year warranty
More About this Item
Round bottom flasks require a cork or rubber ring to stand, or a neck clamp attached to a laboratory support rod
1 Year Warranty
Specifications
  • Capacity (mL)500
  • Flask TypeRound-bottom
  • Flask Joint Size24/40
  • Condenser Joint Size24/40
  • DescriptionSoxhlet Round Bottom Flask; 500 mL; 1/Pk