DWK Life Sciences (Kimble) Kimax Soxhlet Flat Bottom Flask; 300 mL; 1/Pk
Representative image only.
DWK Life Sciences (Kimble) Kimax Soxhlet Flat Bottom Flask; 300 mL; 1/Pk
DWK Life Sciences (Kimble) | Mfr: 601500-0424| Item: 9903748
Choose an extraction flask by volume and bottom shape
₹21,000.00INR / Each
₹21,000.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Kimble) Kimax Soxhlet Flat Bottom Flask; 300 mL; 1/Pk
DWK Life Sciences (Kimble) Kimax Soxhlet Flat Bottom Flask; 300 mL; 1/Pk
9903748
₹21,000.00INR / Each
+ =
Total Price 104000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Flat bottom flasks sit easily on flat surfaces
  • 1 year warranty
More About this Item
Round bottom flasks require a cork or rubber ring to stand, or a neck clamp attached to a laboratory support rod
1 Year Warranty
Specifications
  • Capacity (mL)300
  • Flask TypeFlat-bottom
  • Flask Joint Size24/40
  • Condenser Joint Size24/40
  • DescriptionSoxhlet Flat Bottom Flask; 300 mL; 1/Pk