DWK Life Sciences (Kimble) KC45066-25150 Culture Tube, plastic-coated glass, 50 mL, 48/cs

Representative image only.

DWK Life Sciences (Kimble) KC45066-25150 Culture Tube, plastic-coated glass, 50 mL, 48/cs

DWK Life Sciences (Kimble) | Mfr: KC45066-25150| Item: 3457236

₹161,000.00INR / Case of 48

₹161,000.00INR / Case of 48
Product Description

Key Features

  • Class A glass gives uniform thickness

More About this Item

Ultra-clear safety-coated glassware helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware is ideal for chemical storage, transportation, and handling of hazardous chemicals, harmful biologicals, and acids. Glassware has a unique nonslip surface when wet or dry.Manufactured from KG-33 borosilicate glass for maximum heat transfer and superior chemical resistance. Meets ASTM E982, Type VI requirements. Rubber-lined screw caps are included and are unattached. Autoclavable to120°C (250°F).

Specifications
  • AutoclavableYes
  • Tube Capacity (mL)50
  • Tube MaterialGlass
  • Tube ColorClear
  • Tube Bottom ShapeRound
  • Tube Length (mm)150
  • Tube Outside Diameter (OD) (mm)25
  • Cap IncludedYes
  • Cap StyleScrew Cap
  • GraduationsNone
  • DescriptionCultureTube,plastic-coatedglass,50mL,48/cs
Reviews
Q&A