DWK Life Sciences (Kimble) KC26720-2000 Erlenmeyer Flask with Screw Caps, 2000 mL

Representative image only.

DWK Life Sciences (Kimble) KC26720-2000 Erlenmeyer Flask with Screw Caps, 2000 mL

DWK Life Sciences (Kimble) | Mfr: KC26720-2000| Item: 3452446

KimCote™ plastic coating provides added safety

₹31,800.00INR / Case of 2

₹31,800.00INR / Case of 2
Product Description

More About this Item

Ultra-clear safety-coated flasks helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware is ideal for chemical storage, transportation, and handling of hazardous chemicals, harmful biologicals, and acids.
Glassware has a unique nonslip surface when wet or dry. Durable safety coated flasks feature large opening and GL 45 threaded closures for easy pouring and tight sealing. Graduations are in durable white enamel. Blue caps and clear pour rings are included. Autoclavable to120°C (250°F).

Specifications
  • MaterialGlass
  • Capacity (mL)2000
  • Min Graduation (mL)600
  • Max Graduation (mL)2000
  • Graduation Interval (mL)100
  • Cap TypeGL45 thread
  • Overall Height (cm)26.5
  • DescriptionErlenmeyer Flask with Screw Caps, 2000 mL
Reviews
Q&A