DWK Life Sciences (Kimble) HPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 2.0 Liter
Representative image only.
DWK Life Sciences (Kimble) HPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 2.0 Liter
DWK Life Sciences (Kimble) | Mfr: 953930-2002| Item: 0201750
Coated glass bottles prevent photodegradation of light-sensitive mobile phases
₹146,000.00INR / Each
₹146,000.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Kimble) HPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 2.0 Liter
DWK Life Sciences (Kimble) HPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 2.0 Liter
0201750
₹146,000.00INR / Each
+ + + =
Total Price 166400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of durable safety coated glass
  • 1 year warranty
More About this Item
These complete reservoir systems have chemical-resistant fluid / vapor contact surfaces. The cap body is PTFE with a TFE/propylene o-ring and a polypropylene screw collar. Tubing passes through ¨ù-28 fittings. Cap has two 1/8" through holes and one through hole for 1/16¡± OD tubing. Includes one 2 µm PEEK Inlet/Sparge filter, two ETFE fittings, one ETFE plug, and 10 ft of 1/8" OD FEP PTFE tubing.
The ULTRA-WARE Flat Bottom Reservoir is Type I, Class A borosilicate glass with a GL 45-4 screw thread and a plastic safety coating. The plastic coating blocks UV light up to 385 nm.
1 Year Warranty
Specifications
  • End CapEconomy
  • Bottom TypeFlat
  • Total Volume Capacity (mL)2000
  • DescriptionHPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 2.0 Liter