DWK Life Sciences (Kimble) Glass Pennyhead Stoppers, Standard Taper 24/40; 1/Pk
Representative image only.
DWK Life Sciences (Kimble) Glass Pennyhead Stoppers, Standard Taper 24/40; 1/Pk
DWK Life Sciences (Kimble) | Mfr: 851000-2440| Item: 3470714
₹21,100.00INR / Each
₹21,100.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Sizes Standard Taper 10/30 through 19/22 are solid. Sizes Standard Taper 24/40 and larger are hollow.
1 Year Warranty
Specifications
  • Taper Size24/40
  • MaterialGlass
  • DescriptionGlass Pennyhead Stoppers, Standard Taper 24/40; 1/Pk