DWK Life Sciences (Kimble) Glass Babcock Bottle, Milk Test, 18 gram Sample; 1/Pack
Representative image only.
DWK Life Sciences (Kimble) Glass Babcock Bottle, Milk Test, 18 gram Sample; 1/Pack
DWK Life Sciences (Kimble) | Mfr: 1003S-8| Item: 3459006
₹10,000.00INR / Each
₹10,000.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Kimble) Glass Babcock Bottle, Milk Test, 18 gram Sample; 1/Pack
DWK Life Sciences (Kimble) Glass Babcock Bottle, Milk Test, 18 gram Sample; 1/Pack
3459006
₹10,000.00INR / Each
+ =
Total Price 10000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Easily measure the percentage of butter fat content in your sample with these specialized bottles. The body of each bottle is a reaction chamber, while the graduated neck is a measuring device. Use the Babcock bottles for measuring liquids and small samples. The Paley bottles feature a large opening on the side of the body to easily accommodate larger or viscous samples. Replacement stoppers are available: please see accessories. All bottles measure 6 1/2" H.
Specifications
  • MaterialGlass
  • AutoclavableYes
  • Overall Height (in)6 1/2
  • DescriptionGlass Babcock Bottle, Milk Test, 18 gram Sample; 1/Pack