DWK Life Sciences (Kimble) Conversion Base
Representative image only.
DWK Life Sciences (Kimble) Conversion Base
DWK Life Sciences (Kimble) | Mfr: 953984-0090| Item: 1595888
Modular system to filter HPLC mobile phase directly into reservoir
Call For Price
Call For Price
Product Description
Includes
Supplied with filtration cap, solvent pickup adapter and conical bottom reservoir
Key Features
  • Designed to filter HPLC mobile phase directly into the reservoir using 47mm membrane filter
  • Optional Conversion base allows use of 90mm membrane filter
  • In-line filtration with solvent pickup adapter eliminates traditional pour-and-wait filtration
  • Vacuum degassing is accomplished by closing the HI-VAC® valve
  • Patent No. 5,397,467
  • Recommended for most solvents used for HPLC mobile phase
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
  • DescriptionConversion Base