DWK Life Sciences (Kimble) Bottle Media, 10 L, 1/Cs
Representative image only.
DWK Life Sciences (Kimble) Bottle Media, 10 L, 1/Cs
DWK Life Sciences (Kimble) | Mfr: SP14395-10000| Item: 1595710
Mix, store, and transport culture media, chemicals, and solvents
₹26,000.00INR / Case of 1
₹26,000.00INR / Case of 1
Product Description
More About this Item
These heavy-duty general-purpose Type 1 borosilicate glass bottles have permanent white enamel graduations and marking areas. Each bottle includes an autoclavable (140°C/284°F) linerless blue polypropylene (PP) cap and drip-free pour ring. Maximum temperature: 450°C (840°F). Autoclavable.